Skip to Content
Merck

Skip To

GF53111663

Tungsten

insulated wire, 1m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%

Synonym(s):

Tungsten, W 005810, W

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 356,00

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141747
MDL number:
eCl@ss:
38180901

€ 356,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 531-116-63

resistivity

4.9 μΩ-cm, 20°C

L × diam. × thickness

1 m × 0.075 mm × 0.02 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
267562GF29433348GF55004666
description

insulated wire, 1m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%

description

wire, diam. 1.0 mm, 99.99% trace metals basis

description

wire reel, 25m, diameter 0.75mm, clean, 99.95%

description

wire reel, 500m, diameter 0.125mm, clean, 99.95%

form

wire

form

wire

form

wire

form

wire

assay

99.9%

assay

99.99% trace metals basis

assay

99.95%

assay

99.95%

manufacturer/tradename

Goodfellow 531-116-63

manufacturer/tradename

-

manufacturer/tradename

Goodfellow 294-333-48

manufacturer/tradename

Goodfellow 550-046-66

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters