Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Ships in 14 days - from Aldrich Partner.
| € 128,58 |
250 mg | Ships in 14 days - from Aldrich Partner.
| € 207,65 |
1 g | Ships in 14 days - from Aldrich Partner.
| € 375,39 |
About This Item
Vendor SKU: 41111-100MG
€ 128,58
Ships in 14 days - from Aldrich Partner.
Country of Origin
CN
Inchi Code
1S/2C12H15NO5S.2Na.5H2O/c2*1-5(14)7-10(15)13-8(12(16)17)9(19-11(7)13)6-3-2-4-18-6;;;;;;;/h2*5-7,11,14H,2-4H2,1H3,(H,16,17);;;5*1H2/q;;2*+1;;;;;/p-2/t2*5-,6-,7+,11-;;;;;;;/m11......./s1
Inchi Key
MOGICMVNWAUWMK-HIXRZVNASA-L
Shipping Temperature
Normal
Purity
99%
Physical Form
White to Yellow Solid
Storage temp.
-20 C
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.