Skip to Content
Merck

BDPH9BCC2242

Bide Pharmatech

4-(4-Fluorophenoxy)-2-methylaniline

Synonym(s):

4-(4-fluorophenoxy)-2-methylaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H12O1N1F1
CAS Number:
Molecular Weight:
217.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H12FNO/c1-9-8-12(6-7-13(9)15)16-11-4-2-10(14)3-5-11/h2-8H,15H2,1H3

Inchi Key

XGIQHJPZLVXVKD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library