Skip to Content
Merck

BDPH9BCC2E2C

Bide Pharmatech

Ethyl 1H-1,2,4-triazole-5-carboxylate

Synonym(s):

ethyl 1H-1,2,4-triazole-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H7O2N3
CAS Number:
Molecular Weight:
141.13
MDL number:
Product BDPH9BCC2E2C is not currently sold in your country.


Inchi Code

1S/C5H7N3O2/c1-2-10-5(9)4-6-3-7-8-4/h3H,2H2,1H3,(H,6,7,8)

Inchi Key

DHZYWCBUDKTLGD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library