Skip to Content
Merck

BDPH9BCC2E95

Bide Pharmatech

1-Benzhydryl-3-phenylazetidine

Synonym(s):

1-benzhydryl-3-phenylazetidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H21N1
CAS Number:
Molecular Weight:
299.42
MDL number:
Product BDPH9BCC2E95 is not currently sold in your country.


Inchi Code

1S/C22H21N/c1-4-10-18(11-5-1)21-16-23(17-21)22(19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15,21-22H,16-17H2

Inchi Key

PAXDHGAELYIMBD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 1-Benzhydryl-3-phenylazetidine


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library