Skip to Content
Merck

BDPH9BCC36C8

Bide Pharmatech

5-Iodo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Synonym(s):

5-iodo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7N4I1
CAS Number:
Molecular Weight:
274.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7IN4/c1-12-2-4(8)5-6(9)10-3-11-7(5)12/h2-3H,1H3,(H2,9,10,11)

Inchi Key

VNFSLRASQZPDFQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to light-yellow or gray powder or crystals



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library