Skip to Content

BDPH9BCC394E

Bide Pharmatech

2-((4-Aminophenyl)amino)ethanol

Synonym(s):

2-(4-aminoanilino)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H12O1N2
Molecular Weight:
152.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H12N2O/c9-7-1-3-8(4-2-7)10-5-6-11/h1-4,10-11H,5-6,9H2

Inchi Key

RBUQOUQQUQCSAU-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library