Skip to Content
Merck

BDPH9BCC4429

Bide Pharmatech

Ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate

Synonym(s):

ethyl 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H17O3N1
CAS Number:
Molecular Weight:
223.27
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H17NO3/c1-3-16-11(15)12(2,13)8-9-4-6-10(14)7-5-9/h4-7,14H,3,8,13H2,1-2H3

Inchi Key

LTBRGLHBHNJNHQ-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to off-white powder or crystals



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library