Skip to Content
Merck

BDPH9BCC4A9D

Bide Pharmatech

(4-Amino-2-(isopropylamino)thiazol-5-yl)(phenyl)methanone

Synonym(s):

[4-amino-2-(isopropylamino)-1,3-thiazol-5-yl](phenyl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H15O1N3S1
CAS Number:
Molecular Weight:
261.35
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H15N3OS/c1-8(2)15-13-16-12(14)11(18-13)10(17)9-6-4-3-5-7-9/h3-8H,14H2,1-2H3,(H,15,16)

Inchi Key

WHJIFFVMTRXENN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library