Skip to Content
Merck

BDPH9BCC7FB8

Bide Pharmatech

N-Phenyl-4-(pyridin-3-yl)aniline

Synonym(s):

N-phenyl-4-(pyridin-3-yl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H14N2
CAS Number:
Molecular Weight:
246.31
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H14N2/c1-2-6-16(7-3-1)19-17-10-8-14(9-11-17)15-5-4-12-18-13-15/h1-13,19H

Inchi Key

BXOMRKCAPYLLAC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under N-Phenyl-4-(pyridin-3-yl)aniline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library