Skip to Content
Merck

BDPH9BCCAB34

Bide Pharmatech

3-(Aminomethyl)-6-(trifluoromethyl)pyridine

Synonym(s):

[6-(trifluoromethyl)-3-pyridinyl]methylamine

Select a Size

Change View

About This Item

Linear Formula:
C7H7N2F3
CAS Number:
Molecular Weight:
176.14
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7F3N2/c8-7(9,10)6-2-1-5(3-11)4-12-6/h1-2,4H,3,11H2

Inchi Key

XPXVAYGVYBQKDE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White or Colorless to Yellow Solid or Liquid



Still not finding the right product?


hstatements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library