Skip to Content
Merck

BDPH9BCCACA2

Bide Pharmatech

3,3-Diethoxy-1-propanol

Synonym(s):

3,3-diethoxy-1-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H16O3
CAS Number:
Molecular Weight:
148.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H16O3/c1-3-9-7(5-6-8)10-4-2/h7-8H,3-6H2,1-2H3

Inchi Key

ASERXEZXVIJBRO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: liquid Colour: colourless



Still not finding the right product?

Explore all of our products under 3,3-Diethoxy-1-propanol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library