Inchi Code
1S/2C6H11NO.C2H2O4/c2*1-2-7-3-6(1)4-8-5-6;3-1(4)2(5)6/h2*7H,1-5H2;(H,3,4)(H,5,6)
Inchi Key
DREVFFJHSWBTQS-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
White to Yellow Solid
Still not finding the right product?
Explore all of our products under 2-Oxa-6-azaspiro[3.4]octane hemioxalate
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
