Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C14H11ClO/c1-10-6-8-11(9-7-10)14(16)12-4-2-3-5-13(12)15/h2-9H,1H3
Inchi Key
WIKUXHHNKRKVGJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Sealed in dry, 2-8°C
Still not finding the right product?
Explore all of our products under 2-Chloro-4'-methylbenzophenone
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
