Skip to Content

System maintenance is scheduled for Sunday, October 11, 8:00 AM to 12:00 PM CST (Sunday, 11th October 2026, 03:00 PM to 07:00 PM CET). Quotes, price checks, and order status inquiries may be temporarily unavailable.

Merck

Skip To

BDPH9BCCCBE8

Bide Pharmatech

Ethyl 2-acetoxy-2-(diethoxyphosphoryl)acetate

Synonym(s):

ethyl (acetyloxy)(diethoxyphosphoryl)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C10H19O7P1
CAS Number:
Molecular Weight:
282.23
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H19O7P/c1-5-14-9(12)10(17-8(4)11)18(13,15-6-2)16-7-3/h10H,5-7H2,1-4H3

Inchi Key

QHQNXSICNKWLOC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?


Signal Word

Danger

Pictograms

Exclamation markHealth hazard


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library