Inchi Code
1S/C9H9BrN2O2/c10-7-2-1-3-8(11-7)12-4-6(5-12)9(13)14/h1-3,6H,4-5H2,(H,13,14)
Inchi Key
ZPLNUHDGBKVSEH-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1-(6-Bromopyridin-2-yl)azetidine-3-carboxylic acid
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
