Inchi Code
1S/C4H2Cl2N2O/c5-2-1-7-4(9)3(6)8-2/h1H,(H,7,9)
Inchi Key
XDHJAHNEBHARQO-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Light yellow to yellow powder or crystals
Still not finding the right product?
Explore all of our products under 3,5-Dichloropyrazin-2(1H)-one
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
