Skip to Content
Merck

BDPH9BCCDBBD

Bide Pharmatech

2-Amino-2-(4-chloro-3-fluorophenyl)propan-1-ol

Synonym(s):

2-amino-2-(4-chloro-3-fluorophenyl)-1-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1N1Cl1F1
CAS Number:
Molecular Weight:
203.64
MDL number:
Product BDPH9BCCDBBD is not currently sold in your country.


Inchi Code

1S/C9H11ClFNO/c1-9(12,5-13)6-2-3-7(10)8(11)4-6/h2-4,13H,5,12H2,1H3

Inchi Key

PHAQHIOVPLVAII-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library