Skip to Content
Merck

BDPH9BCCE968

Bide Pharmatech

3,3'-Disulfanediylbis(propan-1-ol)

Synonym(s):

3,3'-disulfanediylbis(propan-1-ol)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H14O2S2
CAS Number:
Molecular Weight:
182.31
MDL number:
Product BDPH9BCCE968 is not currently sold in your country.


Inchi Code

1S/C6H14O2S2/c7-3-1-5-9-10-6-2-4-8/h7-8H,1-6H2

Inchi Key

RXURJOBHVPLGQU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?

Explore all of our products under 3,3'-Disulfanediylbis(propan-1-ol)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library