Skip to Content

BDPH9BCD0D99

Bide Pharmatech

4-Methyl-3-phenylpyridine

Synonym(s):

4-methyl-3-phenylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H11N1
CAS Number:
Molecular Weight:
169.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H11N/c1-10-7-8-13-9-12(10)11-5-3-2-4-6-11/h2-9H,1H3

Inchi Key

YDJITZMUZXZCBI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 4-Methyl-3-phenylpyridine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark

hstatements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library