Skip to Content

BDPH9BCD0EE0

Bide Pharmatech

Decahydropyrrolo[3,4-b]pyrrolizine

Synonym(s):

decahydropyrrolo[3,4-b]pyrrolizine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H16N2
CAS Number:
Molecular Weight:
152.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H16N2/c1-2-8-4-7-5-10-6-9(7)11(8)3-1/h7-10H,1-6H2

Inchi Key

SYZQBOYLQBDNLX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Decahydropyrrolo[3,4-b]pyrrolizine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library