Skip to Content
Merck

BDPH9BCD2010

Bide Pharmatech

5-Bromo-1H-benzo[d]imidazole

Synonym(s):

5-bromo-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5N2Br1
CAS Number:
Molecular Weight:
197.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5BrN2/c8-5-1-2-6-7(3-5)10-4-9-6/h1-4H,(H,9,10)

Inchi Key

GEDVWGDBMPJNEV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow to brown powder or crytals

Melting Point

130-134C



Still not finding the right product?

Explore all of our products under 5-Bromo-1H-benzo[d]imidazole


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library