Skip to Content
Merck

BDPH9BCD2140

Bide Pharmatech

(5-Bromo-6-methylpyridin-2-yl)methanol

Synonym(s):

(5-bromo-6-methyl-2-pyridinyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O1N1Br1
CAS Number:
Molecular Weight:
202.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8BrNO/c1-5-7(8)3-2-6(4-10)9-5/h2-3,10H,4H2,1H3

Inchi Key

JUUUESPVQDOIBC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library