Skip to Content
Merck

BDPH9BCD234A

Bide Pharmatech

Hexadecane

Synonym(s):

hexadecane

Select a Size

Change View

About This Item

Linear Formula:
C16H34
CAS Number:
Molecular Weight:
226.45
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

Inchi Key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to white Liquid or Solid

Melting Point

18C at mmHg



Still not finding the right product?

Explore all of our products under Hexadecane


Precautionary Statements

Signal Word

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library