Skip to Content
Merck

BDPH9BCD262E

Bide Pharmatech

4-Cyano-N-hydroxybenzimidamide

Synonym(s):

4-cyano-N-hydroxybenzenecarboximidamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O1N3
CAS Number:
Molecular Weight:
161.16
MDL number:
Product BDPH9BCD262E is not currently sold in your country.


Inchi Code

1S/C8H7N3O/c9-5-6-1-3-7(4-2-6)8(10)11-12/h1-4,12H,(H2,10,11)

Inchi Key

ZLLAUULXYBIJBX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Cyano-N-hydroxybenzimidamide


hstatements

signalword

Danger

pictograms

Skull and crossbonesEnvironment


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library