Skip to Content
Merck

BDPH9BCD2C4E

Bide Pharmatech

5-Bromo-2-ethoxy-3-methylpyridine

Synonym(s):

5-bromo-2-ethoxy-3-methylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O1N1Br1
CAS Number:
Molecular Weight:
216.08
MDL number:
Product BDPH9BCD2C4E is not currently sold in your country.


Inchi Code

1S/C8H10BrNO/c1-3-11-8-6(2)4-7(9)5-10-8/h4-5H,3H2,1-2H3

Inchi Key

KJXFWKLSNMFKSK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or viscous liquid



Still not finding the right product?

Explore all of our products under 5-Bromo-2-ethoxy-3-methylpyridine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library