Skip to Content
Merck

BDPH9BCD2DFD

Bide Pharmatech

1-(Bromomethyl)-1-methylcyclohexane

Synonym(s):

1-(bromomethyl)-1-methylcyclohexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCD2DFD is not currently sold in your country.

About This Item

Linear Formula:
C8H15Br1
CAS Number:
Molecular Weight:
191.11
MDL number:
Product BDPH9BCD2DFD is not currently sold in your country.

Inchi Code

1S/C8H15Br/c1-8(7-9)5-3-2-4-6-8/h2-7H2,1H3

Inchi Key

QGSSEDLIZIGPGG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library