Skip to Content
Merck

BDPH9BCD2F13

Bide Pharmatech

Methyl 2-(thiophen-3-yl)acetate

Synonym(s):

methyl 3-thienylacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O2S1
CAS Number:
Molecular Weight:
156.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8O2S/c1-9-7(8)4-6-2-3-10-5-6/h2-3,5H,4H2,1H3

Inchi Key

RZGRWVULDSXQSM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Methyl 2-(thiophen-3-yl)acetate


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library