Skip to Content
Merck

BDPH9BCD2F72

Bide Pharmatech

4-Ethynylpicolinaldehyde

Synonym(s):

4-ethynyl-2-pyridinecarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O1N1
CAS Number:
Molecular Weight:
131.13
MDL number:
Product BDPH9BCD2F72 is not currently sold in your country.


Inchi Code

1S/C8H5NO/c1-2-7-3-4-9-8(5-7)6-10/h1,3-6H

Inchi Key

JZXMFQRDHALHMN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?

Explore all of our products under 4-Ethynylpicolinaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library