Skip to Content
Merck

BDPH9BCD38D2

Bide Pharmatech

1-(4-(Dimethylphosphoryl)phenyl)ethan-1-one

Synonym(s):

1-(4-(dimethylphosphoryl)phenyl)ethan-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O2P1
CAS Number:
Molecular Weight:
196.19
MDL number:
Product BDPH9BCD38D2 is not currently sold in your country.


Inchi Code

1S/C10H13O2P/c1-8(11)9-4-6-10(7-5-9)13(2,3)12/h4-7H,1-3H3

Inchi Key

BEJXQFTUCNGGEA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library