Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H10FN/c10-8-4-2-1-3-7(8)9(11)5-6-9/h1-4H,5-6,11H2
Inchi Key
PMUHZJKXCBFDEV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
96%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 1-(2-Fluorophenyl)cyclopropanamine
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
