Skip to Content
Merck

BDPH9BCD43B0

Bide Pharmatech

(1-Ethyl-1H-pyrazol-4-yl)methanamine

Synonym(s):

(1-ethyl-1H-pyrazol-4-yl)methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H11N3
CAS Number:
Molecular Weight:
125.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H11N3/c1-2-9-5-6(3-7)4-8-9/h4-5H,2-3,7H2,1H3

Inchi Key

MIAQYVFWXDEYAK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?


hstatements

Signal Word

Danger

Pictograms

CorrosionExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library