Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H8BrN/c1-7-8(5-6-11)3-2-4-9(7)10/h2-4H,5H2,1H3
Inchi Key
VBNCKBSHNGLORC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Pale-yellow to Yellow Solid
Still not finding the right product?
Explore all of our products under 2-(3-Bromo-2-methylphenyl)acetonitrile
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
