Product BDPH9BCD4ECE is not currently sold in your country.
Inchi Code
1S/C5H4Br2N2/c6-1-5-8-2-4(7)3-9-5/h2-3H,1H2
Inchi Key
BMXJFKLSEGBBSJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
White to yellow powder or crystals
Still not finding the right product?
Explore all of our products under 5-Bromo-2-(bromomethyl)pyrimidine
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

