Skip to Content
Merck

BDPH9BCD5788

Bide Pharmatech

6-Bromo-4-methylpyridin-2-ol

Synonym(s):

6-bromo-4-methylpyridin-2-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N1Br1
CAS Number:
Molecular Weight:
188.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6BrNO/c1-4-2-5(7)8-6(9)3-4/h2-3H,1H3,(H,8,9)

Inchi Key

TYGHHXCOTIUZQO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Bromo-4-methylpyridin-2-ol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library