Skip to Content
Merck

BDPH9BCD5ABF

Bide Pharmatech

2-Bromo-6-(methylthio)pyridine

Synonym(s):

2-bromo-6-(methylsulfanyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6N1Br1S1
CAS Number:
Molecular Weight:
204.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6BrNS/c1-9-6-4-2-3-5(7)8-6/h2-4H,1H3

Inchi Key

CWFMGEWBDIDVPZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to light-yellow liquid



Still not finding the right product?

Explore all of our products under 2-Bromo-6-(methylthio)pyridine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library