Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H9BrO2/c1-12-9(11)6-7-2-4-8(10)5-3-7/h2-5H,6H2,1H3
Inchi Key
QHJOWSXZDCTNQX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Colorless to Pale-yellow to Yellow-brown Liquid
Still not finding the right product?
Explore all of our products under Methyl 2-(4-bromophenyl)acetate
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
