Skip to Content
Merck

BDPH9BCD78D1

Bide Pharmatech

2-(2-Chloro-6-fluorophenyl)-2,2-difluoroacetic acid

Synonym(s):

2-(2-chloro-6-fluorophenyl)-2,2-difluoroacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H4O2Cl1F3
CAS Number:
Molecular Weight:
224.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H4ClF3O2/c9-4-2-1-3-5(10)6(4)8(11,12)7(13)14/h1-3H,(H,13,14)

Inchi Key

JHTZCAVROIRKHO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library