Inchi Code
1S/C8H6ClIO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key
LLLOGZBHLFGNSY-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 5-Chloro-4-iodo-2-methoxybenzoic acid
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
