Skip to Content
Merck

BDPH9BCD85C7

Bide Pharmatech

4-Chloro-2-(ethylthio)quinazoline

Synonym(s):

4-chloro-2-(ethylsulfanyl)quinazoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9N2Cl1S1
CAS Number:
Molecular Weight:
224.71
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9ClN2S/c1-2-14-10-12-8-6-4-3-5-7(8)9(11)13-10/h3-6H,2H2,1H3

Inchi Key

YXMGMRFURWMKDY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Chloro-2-(ethylthio)quinazoline


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library