Skip to Content
Merck

BDPH9BCD8A4E

Bide Pharmatech

2-Methyl-1-pyrroline

Synonym(s):

5-methyl-2,3-dihydro-1H-pyrrole

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCD8A4E is not currently sold in your country.

About This Item

Linear Formula:
C5H9N1
CAS Number:
Molecular Weight:
83.13
MDL number:
Product BDPH9BCD8A4E is not currently sold in your country.

Inchi Code

1S/C5H9N/c1-5-3-2-4-6-5/h3,6H,2,4H2,1H3

Inchi Key

MUUWQYQRBFVTIB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

pictograms

FlameExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library