Inchi Code
1S/C5H6BrN3O2/c1-11-5(10)3-2(6)4(7)9-8-3/h1H3,(H3,7,8,9)
Inchi Key
VQCLUHFFNGRFCC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under Methyl 3-amino-4-bromo-1H-pyrazole-5-carboxylate
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
