Skip to Content
Merck

BDPH9BCD910C

Bide Pharmatech

4-Bromo-2-fluoro-5-methoxyaniline

Synonym(s):

4-bromo-2-fluoro-5-methoxyaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O1N1Br1F1
CAS Number:
Molecular Weight:
220.04
MDL number:
Product BDPH9BCD910C is not currently sold in your country.


Inchi Code

1S/C7H7BrFNO/c1-11-7-3-6(10)5(9)2-4(7)8/h2-3H,10H2,1H3

Inchi Key

JGQVADKRUVUCGR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Bromo-2-fluoro-5-methoxyaniline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library