Skip to Content
Merck

BDPH9BCD91EB

Bide Pharmatech

4-Amino-2,6-dichloronicotinic acid

Synonym(s):

4-amino-2,6-dichloronicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4O2N2Cl2
CAS Number:
Molecular Weight:
207.02
MDL number:
Product BDPH9BCD91EB is not currently sold in your country.


Inchi Code

1S/C6H4Cl2N2O2/c7-3-1-2(9)4(6(11)12)5(8)10-3/h1H,(H2,9,10)(H,11,12)

Inchi Key

VOVZKEQDQUXIDG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Amino-2,6-dichloronicotinic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library