Skip to Content
Merck

BDPH9BCD9DFD

Bide Pharmatech

6-(tert-Butoxy)pyridin-3-amine

Synonym(s):

6-tert-butoxy-3-pyridinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCD9DFD is not currently sold in your country.

About This Item

Linear Formula:
C9H14O1N2
CAS Number:
Molecular Weight:
166.22
MDL number:
Product BDPH9BCD9DFD is not currently sold in your country.

Inchi Code

1S/C9H14N2O/c1-9(2,3)12-8-5-4-7(10)6-11-8/h4-6H,10H2,1-3H3

Inchi Key

HRLHJWKMOMDDND-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or liquid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library