Skip to Content
Merck

BDPH9BCDA0FA

Bide Pharmatech

4-(Benzyloxy)-3-(trifluoromethyl)aniline

Synonym(s):

4-(benzyloxy)-3-(trifluoromethyl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H12O1N1F3
CAS Number:
Molecular Weight:
267.25
MDL number:
Product BDPH9BCDA0FA is not currently sold in your country.


Inchi Code

1S/C14H12F3NO/c15-14(16,17)12-8-11(18)6-7-13(12)19-9-10-4-2-1-3-5-10/h1-8H,9,18H2

Inchi Key

UULCQZNHBGUYCE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library