Skip to Content
Merck

BDPH9BCDA32E

Bide Pharmatech

4-Chloro-5-methylpicolinic acid

Synonym(s):

4-chloro-5-methyl-2-pyridinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H6O2N1Cl1
CAS Number:
Molecular Weight:
171.58
MDL number:
Product BDPH9BCDA32E is not currently sold in your country.


Inchi Code

1S/C7H6ClNO2/c1-4-3-9-6(7(10)11)2-5(4)8/h2-3H,1H3,(H,10,11)

Inchi Key

IIYOXHJTMDXVMM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?

Explore all of our products under 4-Chloro-5-methylpicolinic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library