Skip to Content

BDPH9BCDA52B

Bide Pharmatech

(4-Ethoxy-2-fluorophenyl)methanol

Synonym(s):

(4-ethoxy-2-fluorophenyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O2F1
CAS Number:
Molecular Weight:
170.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11FO2/c1-2-12-8-4-3-7(6-11)9(10)5-8/h3-5,11H,2,6H2,1H3

Inchi Key

QOYFXEFXXGGUIF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: liquid



Still not finding the right product?

Explore all of our products under (4-Ethoxy-2-fluorophenyl)methanol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library