Inchi Code
1S/C8H9BF2O3/c1-2-14-8-4-6(10)5(9(12)13)3-7(8)11/h3-4,12-13H,2H2,1H3
Inchi Key
MCHGZBKHWUZZTJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (4-Ethoxy-2,5-difluorophenyl)boronic acid
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
