Skip to Content

BDPH9BCDB383

Bide Pharmatech

2-(Furan-2-yl)pyrimidine-5-carboxylic acid

Synonym(s):

2-(2-furyl)-5-pyrimidinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O3N2
CAS Number:
Molecular Weight:
190.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6N2O3/c12-9(13)6-4-10-8(11-5-6)7-2-1-3-14-7/h1-5H,(H,12,13)

Inchi Key

GVRCLBMNNYVPKE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library